This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GM11541 SIRNA Assays ›
Gene Symbol:
Gm11541
Gene Name:
predicted gene 11541
Gene Aliases:
OTTMUSG00000002038
Chromosome Location:
Chr.11: 94694498 - 94704499 on Build GRCm38
UniGene ID:
Mm.342314
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 217824

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm11541
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007584.2 NP_001007585.1 1, 2 422
GenBank mRNA AK082500.1 - 1, 2 423
BC147833.1 - 1, 2 423 9087550
BC147838.1 - 1, 2 423 9087555

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-lrzm9:80/100.66.79.177:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0