siRNA Details
| Gene Symbol: Ggps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007626.2 | NP_001007627.1 | 4 | 1061 | ||
| XM_008771699.1 | XP_008769921.1 | 3 | 986 | |||
| XM_008771700.1 | XP_008769922.1 | 3 | 1008 | |||
| GenBank mRNA | AB118238.1 | - | 4 | 1047 | ||
| AB118239.1 | - | 4 | 1047 | |||
| AB118240.1 | - | 4 | 1047 | |||
| BC090327.1 | - | 4 | 1061 | 7125199 | ||

Ggps1
Genomic Map