siRNA Details
| Gene Symbol: Phospho2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007642.1 | NP_001007643.1 | 4 | 1284 | ||
| GenBank mRNA | BC081926.1 | - | 4 | 1284 | 7113082 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phospho2| Gene Symbol: Phospho2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007642.1 | NP_001007643.1 | 4 | 1284 | ||
| GenBank mRNA | BC081926.1 | - | 4 | 1284 | 7113082 | |