This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other VWA9 SIRNA Assays ›
Gene Symbol:
Vwa9
Gene Name:
von Willebrand factor A domain containing 9
Gene Aliases:
RGD1359616
Chromosome Location:
Chr.8: 70446159 - 70471140 on Build Rnor_6.0
UniGene ID:
Rn.9525
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 218022

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vwa9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007663.1 NP_001007664.1 11, 12 1679
XM_008766238.1 XP_008764460.1 12, 13 1981
XM_008766239.1 XP_008764461.1 13, 14 1933
GenBank mRNA BC082006.1 - 11, 12 1679 7131624
FQ231531.1 - 7, 8 759

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-ftbch:80/100.66.79.106:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0