This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMEM206 SIRNA Assays ›
Gene Symbol:
Tmem206
Gene Name:
transmembrane protein 206
Gene Aliases:
RGD1359339
Chromosome Location:
Chr.13: 110006975 - 110032790 on Build Rnor_6.0
UniGene ID:
Rn.18267
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 218065

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem206
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007679.1 NP_001007680.1 4 524
XM_006250481.2 XP_006250543.1 3 348
XM_008769855.1 XP_008768077.1 4 559
GenBank mRNA BC082061.1 - 4 524 7132483
FQ225543.1 - 4 533
FQ228821.1 - 4 566
FQ233431.1 - 4 532

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025864, AK031270, AK049641, AK012107, AK044482, and others.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline