This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FAM213A SIRNA Assays ›
Gene Symbol:
Fam213a
Gene Name:
family with sequence similarity 213, member A
Gene Aliases:
RGD1305689, Tmem254
Chromosome Location:
Chr.16: 3817402 - 3821270 on Build Rnor_6.0
UniGene ID:
Rn.35882
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 218400

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam213a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008297.1 NP_001008298.1 4 569
XM_006252573.1 XP_006252635.1 4 517
GenBank mRNA BC086323.1 - 4 569 7304524
FQ213064.1 - 403
FQ220194.1 - 4 396

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline