This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other COL4A4 SIRNA Assays ›
Gene Symbol:
Col4a4
Gene Name:
collagen, type IV, alpha 4
Gene Aliases:
-
Chromosome Location:
Chr.9: 88314763 - 88357183 on Build Rnor_6.0
UniGene ID:
Rn.22075
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 218499

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Col4a4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008332.1 NP_001008333.1 7 939
GenBank mRNA BC086403.1 - 7 939 7304199

Back To Top

More Information


Panther Classification:

Molecular Function -

extracellular matrix structural protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
glomerular basement membrane development
molecular_function
extracellular matrix structural constituent

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nlplw:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline