This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RGD1309594 SIRNA Assays ›
Gene Symbol:
RGD1309594
Gene Name:
similar to RIKEN cDNA 1810043G02; DNA segment, Chr 10, Johns Hopkins University 13, expressed
Gene Aliases:
-
Chromosome Location:
Chr.20: 11416966 - 11424301 on Build Rnor_6.0
UniGene ID:
Rn.128896
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 218555

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RGD1309594
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008351.1 NP_001008352.1 7 1643
XM_006256252.2 XP_006256314.1 7 1941
GenBank mRNA BC085944.1 - 7 1643 7127440

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026431, AK075794, AK165293, AK003097, AK213576, and others.

Gene Ontology Categories:

Function(s)

cytoskeleton organization
smoothened signaling pathway
regulation of cell shape
cilium morphogenesis

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-5kb8h:80/100.66.73.193:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0