This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TRAPPC3 SIRNA Assays ›
Gene Symbol:
Trappc3
Gene Name:
trafficking protein particle complex 3
Gene Aliases:
-
Chromosome Location:
Chr.5: 144281720 - 144295306 on Build Rnor_6.0
UniGene ID:
Rn.925
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 218625

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Trappc3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008376.1 NP_001008377.1 2 127
GenBank mRNA BC086377.1 - 2 127 7301917
FQ214189.1 - 2 183

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013718, AF041433, BC003736, AK168518, AK151747.

Gene Ontology Categories:

Function(s)

ER to Golgi vesicle-mediated transport

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-ct9lx:80/100.66.79.220:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0