This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GM5595 SIRNA Assays ›
Gene Symbol:
Gm5595 Zfp936
Gene Name:
predicted gene 5595
zinc finger protein 936
Gene Aliases:
EG434179, mszf88
EG435970, EG668620, Gm9272, I1C0022H11Rik
Chromosome Location:
Chr.7: 42660105 - 42692732 on Build GRCm38
Chromosome Location:
Chr.7: 43177588 - 43192109 on Build GRCm38
UniGene ID:
Mm.284947
Mm.388871
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID 218706

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm5595
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008427.1 NP_001008427.1 4 1328
GenBank mRNA AB010373.1 - 1 44
BC068175.1 - 4 1328 30550329
Gene Symbol: Zfp936
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034893.1 NP_001030065.1 4 590
GenBank mRNA AK145615.1 - 4 590

Back To Top

More Information


Panther Classification:

Molecular Function -

DNA-binding transcription factor C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-2hkxv:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline