This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CCIN SIRNA Assays ›
Gene Symbol:
Ccin
Gene Name:
calicin
Gene Aliases:
4933417A14Rik
Chromosome Location:
Chr.4: 43983504 - 43985533 on Build GRCm38
UniGene ID:
Mm.348215
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 219940

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ccin
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002787.2 NP_001002787.1 1 1559
GenBank mRNA AK030197.1 - 1 1562
BC150949.1 - 1 1546 9087860
BC150952.1 - 1 1546 9087863

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001005904, BC083832.

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
spermatogenesis
biological_process
cell differentiation
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nlc25:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline