siRNA Details
| Gene Symbol: Fam170a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004061.1 | NP_001004061.1 | 3 | 791 | ||
| GenBank mRNA | AY692448.1 | - | 2 | 647 | ||
| AY692449.1 | - | 3 | 791 | |||
| BC147592.1 | - | 3 | 789 | 9056582 | ||
| BC147595.1 | - | 3 | 789 | 9056585 | ||
| BC172108.1 | - | 3 | 789 | |||
| BC172111.1 | - | 3 | 713 | |||
| BC172113.1 | - | 3 | 713 | |||

Fam170a
Genomic Map