siRNA Details
| Gene Symbol: Ddx56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004211.1 | NP_001004211.1 | 6, 7 | 1113 | ||
| XM_006251437.2 | XP_006251499.1 | 7, 8 | 1048 | |||
| GenBank mRNA | BC078919.1 | - | 6, 7 | 1113 | 7096512 | |
| FQ212430.1 | - | 1034 | ||||

Ddx56
Genomic Map