This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMEM106B SIRNA Assays ›
Gene Symbol:
Tmem106b
Gene Name:
transmembrane protein 106B
Gene Aliases:
LRRGT00101
Chromosome Location:
Chr.4: 39517679 - 39534491 on Build Rnor_6.0
UniGene ID:
Rn.106000
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 220647

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem106b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004267.1 NP_001004267.1 5, 6 572
GenBank mRNA AY387087.1 - 775
BC079127.1 - 5, 6 572 7127112

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_027992, BC050246, AK009646.

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
lysosome localization
dendrite morphogenesis
molecular_function
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-b4tm8:80/100.66.75.155:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline