This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MGC95208 SIRNA Assays ›
Gene Symbol:
MGC95208 LOC100362724
Gene Name:
similar to 4930453N24Rik protein
hypothetical protein LOC100362724
Gene Aliases:
-
-
Chromosome Location:
Chr.11: 1814294 - 1818942 on Build Rnor_6.0
Chromosome Location:
Chr.X: 20563056 - 20564380 on Build Rnor_6.0
UniGene ID:
Rn.19101
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID 221084

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MGC95208
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005552.1 NP_001005552.1 3 1072
GenBank mRNA BC082055.1 - 3 1072 7132244
BC091244.1 - 3 1067 7365330
Gene Symbol: LOC100362724
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003752027.2 XP_003752075.1 1 1062

Back To Top

More Information


Gene Ontology Categories:

Function(s)

apoptotic process

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-9w28n:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline