siRNA Details
| Gene Symbol: Pi4k2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005883.1 | NP_001005883.1 | 5 | 883 | ||
| GenBank mRNA | BC083668.1 | - | 5 | 883 | 7135573 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pi4k2b| Gene Symbol: Pi4k2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005883.1 | NP_001005883.1 | 5 | 883 | ||
| GenBank mRNA | BC083668.1 | - | 5 | 883 | 7135573 | |