siRNA Details
| Gene Symbol: ZNF473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017026574.1 | XP_016882063.1 | 3 | 5151 | ||
| GenBank mRNA | AB032967.1 | - | 5 | 3908 | ||
| AL080143.1 | - | 1 | 2217 | |||
| BX648093.1 | - | 5 | 3952 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF473| Gene Symbol: ZNF473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017026574.1 | XP_016882063.1 | 3 | 5151 | ||
| GenBank mRNA | AB032967.1 | - | 5 | 3908 | ||
| AL080143.1 | - | 1 | 2217 | |||
| BX648093.1 | - | 5 | 3952 | |||