This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GLTSCR1L SIRNA Assays ›
Gene Symbol:
GLTSCR1L
Gene Name:
GLTSCR1 like
Gene Aliases:
KIAA0240
Chromosome Location:
Chr.6: 42746958 - 42868560 on Build GRCh38
UniGene ID:
Hs.658033
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 221362

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GLTSCR1L
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318819.1 NP_001305748.1 7 2036
NM_015349.2 NP_056164.1 5 1769
GenBank mRNA AK091434.1 - 5 1661
AK093526.1 - 5 2380
AK294706.1 - 5 1661
AL833540.1 - 5 1769
BC130302.1 - 5 1692 40146333
CR627378.1 - 6 1734
D87077.2 - 8 1933

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4zngx:80/100.66.79.65:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline