This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other IGFL2 SIRNA Assays ›
Gene Symbol:
IGFL2
Gene Name:
IGF like family member 2
Gene Aliases:
UNQ645, VPRI645
Chromosome Location:
Chr.19: 46078513 - 46195097 on Build GRCh38
UniGene ID:
Hs.99376
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 222631

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: IGFL2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002915.2 NP_001002915.2 5 805
NM_001135113.1 NP_001128585.1 4 517
XM_011526501.1 XP_011524803.1 5 3344
XM_017026335.1 XP_016881824.1 4 1193
GenBank mRNA AK093330.1 - 3 1831
AK125754.1 - 5 1663
AY358692.1 - 4 517
AY672112.1 - 5 805
BC130490.1 - 4 606 40146521
BC130492.1 - 4 606 40146523

Back To Top

More Information


Panther Classification:

Molecular Function -

growth factor intercellular signal molecule

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-smrv7:80/100.66.72.156:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0