This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OLR1570 SIRNA Assays ›
Gene Symbol:
Olr1570
Gene Name:
olfactory receptor 1570
Gene Aliases:
ratchr11-83815769-83816710_ORF
Chromosome Location:
Chr.11: 85398979 - 85399920 on Build Rnor_6.0
UniGene ID:
Rn.142226
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 222769

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olr1570
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000041.1 NP_001000041.1 1 821

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_146451, BC145676.

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0