siRNA Details
| Gene Symbol: DNAJB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001394.3 | NP_001001394.1 | 1 | 1116 | ||
| GenBank mRNA | AY083665.1 | - | 1 | 1116 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNAJB3| Gene Symbol: DNAJB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001394.3 | NP_001001394.1 | 1 | 1116 | ||
| GenBank mRNA | AY083665.1 | - | 1 | 1116 | ||