siRNA Details
| Gene Symbol: MON1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286639.1 | NP_001273568.1 | 2 | 436 | ||
| NM_014940.3 | NP_055755.1 | 2 | 436 | |||
| GenBank mRNA | AB020679.1 | - | 2 | 120 | ||
| AF442486.1 | - | 1 | 100 | |||
| AK023374.1 | - | 2 | 386 | |||
| AK023896.1 | - | 2 | 164 | |||
| AK296467.1 | - | 2 | 163 | |||
| BC024277.1 | - | 2 | 416 | 4843859 | ||
| CF125013.1 | - | 2 | 393 | |||
| DC348034.1 | - | 2 | 436 | |||

MON1B
Genomic Map