This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GLIPR2 SIRNA Assays ›
Gene Symbol:
Glipr2
Gene Name:
GLI pathogenesis-related 2
Gene Aliases:
5730414A08Rik, C77180, GAPR-1
Chromosome Location:
Chr.4: 43957702 - 43979118 on Build GRCm38
UniGene ID:
Mm.22213
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 223426

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Glipr2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027450.3 NP_081726.1 5 1030
GenBank mRNA AK017557.1 - 5 1007
AK141459.1 - 5 1021
AK154433.1 - 5 1008
AK167595.1 - 5 1030
BC031750.1 - 5 1030 4037002

Back To Top

More Information


Panther Classification:

Molecular Function -

defense/immunity protein

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of epithelial cell migration
positive regulation of epithelial to mesenchymal transition
positive regulation of ERK1 and ERK2 cascade
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-rlpqk:80/100.66.76.251:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline