This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other AMTN SIRNA Assays ›
Gene Symbol:
AMTN
Gene Name:
amelotin
Gene Aliases:
UNQ689
Chromosome Location:
Chr.4: 70518572 - 70532743 on Build GRCh38
UniGene ID:
Hs.453069
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 223765

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: AMTN
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286731.1 NP_001273660.1 6, 7 433
NM_212557.3 NP_997722.1 6, 7 436
GenBank mRNA AB593161.1 - 6, 7 435
AY358528.1 - 6, 7 427
BC121812.1 - 6, 7 404 40109020
BC121817.1 - 6, 7 407 40109016

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
biomineral tissue development
odontogenesis of dentin-containing tooth
positive regulation of biomineral tissue development
positive regulation of enamel mineralization
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nlplw:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline