This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ST8SIA6 SIRNA Assays ›
Gene Symbol:
St8sia6
Gene Name:
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 6
Gene Aliases:
Siat8f
Chromosome Location:
Chr.17: 80960785 - 81002830 on Build Rnor_6.0
UniGene ID:
Rn.104457
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 223811

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: St8sia6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_213624.1 NP_998789.1 473
GenBank mRNA AJ699423.1 - 473

Back To Top

More Information


Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

ganglioside biosynthetic process
protein O-linked glycosylation
glycoprotein metabolic process
glycolipid biosynthetic process
oligosaccharide metabolic process
carbohydrate biosynthetic process
sialylation
sialyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0