siRNA Details
| Gene Symbol: Fam19a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182808.3 | NP_877960.1 | 5 | 1645 | ||
| GenBank mRNA | AK049880.1 | - | 4 | 1536 | ||
| AK050001.1 | - | 5 | 1646 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam19a1| Gene Symbol: Fam19a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182808.3 | NP_877960.1 | 5 | 1645 | ||
| GenBank mRNA | AK049880.1 | - | 4 | 1536 | ||
| AK050001.1 | - | 5 | 1646 | |||