This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CHDH SIRNA Assays ›
Gene Symbol:
Chdh
Gene Name:
choline dehydrogenase
Gene Aliases:
CDH, CHD
Chromosome Location:
Chr.16: 6078086 - 6109386 on Build Rnor_6.0
UniGene ID:
Rn.22857
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 224053

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Chdh
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198731.2 NP_942026.1 8 2209
XM_006252578.2 XP_006252640.1 8 2274
XM_006252579.2 XP_006252641.1 8 2228
XM_006252580.2 XP_006252642.1 8 2034
XM_008770984.1 XP_008769206.1 8 2577
XM_008770985.1 XP_008769207.1 9 2666
GenBank mRNA BC085787.1 - 8 2209 7108667
X94769.1 - 7 1484

Back To Top

More Information


Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

glycine betaine biosynthetic process from choline
oxidation-reduction process
choline dehydrogenase activity
flavin adenine dinucleotide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-stzfq:80/100.66.77.128:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0