This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ABR SIRNA Assays ›
Gene Symbol:
Abr
Gene Name:
active BCR-related gene
Gene Aliases:
-
Chromosome Location:
Chr.11: 76416732 - 76577739 on Build GRCm38
UniGene ID:
Mm.258939
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 224090

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Abr
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291186.1 NP_001278115.1 18 3159
NM_198018.2 NP_932135.1 22 3934
NM_198894.2 NP_942597.1 23 3820
NM_198895.2 NP_942598.1 23 3941
GenBank mRNA AK134993.1 - 18 3147
BC043673.1 - 8 1913
BC056385.1 - 18 3148 6854368
BC058708.1 - 22 3934 6839300
BC059064.1 - 18 3143 6825281

Back To Top

More Information


Panther Classification:

Molecular Function -

G-protein modulator

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
brain development
actin cytoskeleton organization
negative regulation of cell migration
response to lipopolysaccharide
regulation of Rho protein signal transduction
intracellular signal transduction
inner ear morphogenesis
negative regulation of neutrophil degranulation
positive regulation of GTPase activity
negative regulation of inflammatory response
positive regulation of phagocytosis
neuromuscular process controlling balance
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
GTPase activator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline