This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF365 SIRNA Assays ›
Gene Symbol:
ZNF365
Gene Name:
zinc finger protein 365
Gene Aliases:
Su48, UAN, ZNF365D
Chromosome Location:
Chr.10: 62373609 - 62672011 on Build GRCh38
UniGene ID:
Hs.22653
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 22446

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF365
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014951.2 NP_055766.2 2 658
NM_199450.2 NP_955522.1 2 658
NM_199451.2 NP_955523.1 2 658
XM_017015937.1 XP_016871426.1 2 1206
GenBank mRNA AB020651.1 - 2 620
AJ505147.1 - 2 431
AJ505148.1 - 2 620
AJ505149.1 - 2 620
AK289515.1 - 2 399
AK289808.1 - 2 423
BC017841.1 - 2 462
BC070073.1 - 2 443
BX647853.1 - 2 451
BX647904.1 - 2 451

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline