This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RRP12 SIRNA Assays ›
Gene Symbol:
RRP12
Gene Name:
ribosomal RNA processing 12 homolog
Gene Aliases:
KIAA0690
Chromosome Location:
Chr.10: 97356701 - 97401370 on Build GRCh38
UniGene ID:
Hs.434251
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 22728

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RRP12
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145114.1 NP_001138586.1 2, 3 513
NM_001284337.1 NP_001271266.1 2, 3 513
NM_015179.3 NP_055994.2 2, 3 513
XM_011539556.2 XP_011537858.1 2, 3 483
GenBank mRNA AB014590.1 - 2, 3 426
AK022496.1 - 2, 3 484
AK296315.1 - 2, 3 377
BC012745.2 - 2, 3 451 4025242
DA227012.1 - 2, 3 483
DC417040.1 - 2, 3 513

Back To Top

More Information


Gene Ontology Categories:

Process(es)

poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mlg7h:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline