This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ARMC8 SIRNA Assays ›
Gene Symbol:
ARMC8
Gene Name:
armadillo repeat containing 8
Gene Aliases:
GID5, HSPC056, S863-2, VID28
Chromosome Location:
Chr.3: 138187248 - 138298389 on Build GRCh38
UniGene ID:
Hs.744868
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 22807

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ARMC8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267041.2 NP_001253970.1 17, 18 1935
NM_001267042.2 NP_001253971.1 17, 18 1827
NM_001282342.1 NP_001269271.1 16, 17 1858
NM_015396.5 NP_056211.2 19, 20 2104
XM_006713564.3 XP_006713627.1 18, 19 2023
XM_006713565.2 XP_006713628.1 19, 20 2064
XM_006713566.3 XP_006713629.1 17, 18 1918
XM_006713567.3 XP_006713630.1 18, 19 2023
XM_011512632.1 XP_011510934.1 18, 19 1971
XM_011512633.1 XP_011510935.1 18, 19 2025
XM_017006095.1 XP_016861584.1 18, 19 1825
XM_017006096.1 XP_016861585.1 17, 18 1918
XM_017006097.1 XP_016861586.1 17, 18 1771
XM_017006098.1 XP_016861587.1 16, 17 1717
XM_017006099.1 XP_016861588.1 17, 18 1808
GenBank mRNA AK001721.1 - 17, 18 1665
AK001882.1 - 10, 11 1239
AK296757.1 - 17, 18 1558
AK296837.1 - 17, 18 1635
AK299229.1 - 13, 14 1296
AK299769.1 - 16, 17 1578
AK304558.1 - 17, 18 1611
AK315932.1 - 13, 14 1301
AL096748.1 - 5, 6 1125
BC032661.1 - 19, 20 1968 5530425
BC041699.1 - 19, 20 1783 5240423

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-fbxsq:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline