This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF451 SIRNA Assays ›
Gene Symbol:
ZNF451
Gene Name:
zinc finger protein 451
Gene Aliases:
COASTER, dJ417I1.1
Chromosome Location:
Chr.6: 57089203 - 57170305 on Build GRCh38
UniGene ID:
Hs.485628
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 23078

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF451
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031623.2 NP_001026794.1 5, 6 656
NM_015555.2 NP_056370.2 5, 6 656
XM_005248994.1 XP_005249051.1 5, 6 676
XM_011514460.1 XP_011512762.1 4, 5 415
XM_011514461.1 XP_011512763.1 3, 4 1031
XM_011514462.2 XP_011512764.1 5, 6 676
GenBank mRNA AB011148.1 - 5, 6 475
AK225806.1 - 5, 6 483
AY055204.1 - 5, 6 657
BC021712.2 - 5, 6 573 4821000
BC098382.1 - 5, 6 499 5295845
BX648081.1 - 5, 6 502
CN314846.1 - 1, 2 59

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
protein sumoylation
negative regulation of transforming growth factor beta receptor signaling pathway
negative regulation of transcription initiation from RNA polymerase II promoter
negative regulation of histone H3-K9 acetylation
RNA polymerase II transcription corepressor activity
transcription factor activity, sequence-specific DNA binding
protein binding
ligase activity
metal ion binding
SUMO ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rdsp8:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline