This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other NSMF SIRNA Assays ›
Gene Symbol:
NSMF
Gene Name:
NMDA receptor synaptonuclear signaling and neuronal migration factor
Gene Aliases:
HH9, NELF
Chromosome Location:
Chr.9: 137447570 - 137459357 on Build GRCh38
UniGene ID:
Hs.455336
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 23164

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NSMF
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130969.1 NP_001124441.1 16 2182
NM_001130970.1 NP_001124442.1 15 2113
NM_001130971.1 NP_001124443.1 14 2107
NM_001178064.1 NP_001171535.1 15 2092
NM_015537.4 NP_056352.3 15 2176
XM_005266061.4 XP_005266118.1 14 2109
XM_005266062.4 XP_005266119.1 13 2040
XM_011518496.2 XP_011516798.1 14 2046
XM_011518497.2 XP_011516799.1 11 1296
XM_017014597.1 XP_016870086.1 10 1565
GenBank mRNA AJ420418.1 - 8 957
AK027474.1 - 13 1447
AK303976.1 - 11 1343
AK316046.1 - 13 1498
AL117660.2 - 11 1204
AL389944.1 - 11 1230
AL389945.1 - 9 2076
AL389946.1 - 7 870
AY255128.1 - 16 2182
AY255129.1 - 15 2176
AY255130.1 - 15 2113
AY255131.1 - 15 2092
AY255132.1 - 15 2418
BC004318.1 - 7 850
BC016862.2 - 11 1223
BC072412.1 - 15 2161 6165009
BC110498.2 - 14 1974 40007676

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein dephosphorylation
regulation of neuron apoptotic process
regulation of neuronal synaptic plasticity
regulation of dendrite morphogenesis
cellular response to amino acid stimulus
cellular response to electrical stimulus
cellular response to gonadotropin stimulus
positive regulation of neuron migration
calcium-dependent protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-5kb8h:80/100.66.73.193:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0