This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RTEL1-TNFRSF6B SIRNA Assays ›
Gene Symbol:
RTEL1-TNFRSF6B RTEL1
Gene Name:
RTEL1-TNFRSF6B readthrough (NMD candidate)
regulator of telomere elongation helicase 1
Gene Aliases:
-
C20orf41, DKCA4, DKCB5, NHL, PFBMFT3, RTEL
Chromosome Location:
Chr.20: 63657810 - 63698698 on Build GRCh38
Chromosome Location:
Chr.20: 63657810 - 63696253 on Build GRCh38
UniGene ID:
Hs.745057
Hs.745057
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID 23311

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RTEL1-TNFRSF6B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_037882.1 29 3595
Gene Symbol: RTEL1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001283009.1 NP_001269938.1 29 3595
NM_001283010.1 NP_001269939.1 28 3316
NM_016434.3 NP_057518.1 29 3595
NM_032957.4 NP_116575.3 29 3667
GenBank mRNA AF217795.1 - 29 3595
AK000485.1 - 5 798
AK299332.1 - 29 3090
AK302508.1 - 29 3186
AK304798.1 - 28 2812
AL080127.1 - 6 648
JX119186.1 - 28 2768

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Panther Classification:

Molecular Function -

DNA helicase

Gene Ontology Categories:

Function(s) Process(es)

telomere maintenance
strand displacement
DNA repair
regulation of double-strand break repair via homologous recombination
replication fork processing
positive regulation of telomere maintenance
DNA duplex unwinding
telomere maintenance in response to DNA damage
telomeric loop disassembly
mitotic telomere maintenance via semi-conservative replication
positive regulation of telomere capping
positive regulation of telomere maintenance via telomere lengthening
negative regulation of t-circle formation
negative regulation of telomere maintenance in response to DNA damage
positive regulation of telomeric loop disassembly
DNA binding
ATP-dependent DNA helicase activity
protein binding
ATP binding
metal ion binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-5xfzg:80/100.66.79.65:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline