This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CFAP36 SIRNA Assays ›
Gene Symbol:
CFAP36 HNRNPM ZNF232
Gene Name:
cilia and flagella associated protein 36
heterogeneous nuclear ribonucleoprotein M
zinc finger protein 232
Gene Aliases:
BARTL1, CCDC104
CEAR, HNRNPM4, HNRPM, HNRPM4, HTGR1, NAGR1, hnRNP M
ZSCAN11
Chromosome Location:
Chr.2: 55519595 - 55545080 on Build GRCh38
Chromosome Location:
Chr.19: 8444575 - 8489118 on Build GRCh38
Chromosome Location:
Chr.17: 5105734 - 5123116 on Build GRCh38
UniGene ID:
Hs.264208
Hs.465808
Hs.279914
Species:
Human
Species Specific ID (Flybase ID):
-
-
-
siRNA ID 235638

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CFAP36
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC001797.1 - 2354
Gene Symbol: HNRNPM
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK309659.1 - 1585
Gene Symbol: ZNF232
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC002852.1 - 2728

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-7wnc4:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline