This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TAOK2 SIRNA Assays ›
Gene Symbol:
TAOK2
Gene Name:
TAO kinase 2
Gene Aliases:
MAP3K17, PSK, PSK1, PSK1-BETA, TAO1, TAO2
Chromosome Location:
Chr.16: 29973641 - 29992261 on Build GRCh38
UniGene ID:
Hs.291623
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 24014

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TAOK2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252043.1 NP_001238972.1 17 3114
NM_016151.3 NP_057235.2 16 3453
XM_011545982.1 XP_011544284.1 16 3700
XM_011545983.2 XP_011544285.1 16 3697
GenBank mRNA AF061943.1 - 16 3394
AL137701.1 - 8 3290
BC051798.1 - 16 3210
BC136653.1 - 16 3138 9020643
BC136655.1 - 16 3138 9020645
BC142663.1 - 16 2793 40148112
BC144344.1 - 17 2799 9052864
BC151221.1 - 16 2793 40148752

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPKK activity
regulation of cell growth
protein targeting to membrane
apoptotic process
response to stress
cellular response to DNA damage stimulus
regulation of cell shape
cell migration
actin cytoskeleton organization
G2 DNA damage checkpoint
positive regulation of stress-activated MAPK cascade
positive regulation of JNK cascade
focal adhesion assembly
stress-activated MAPK cascade
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
ATP binding
mitogen-activated protein kinase kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-v4fgr:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline