This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LRRC37A SIRNA Assays ›
Gene Symbol:
LRRC37A LOC105376839 LRRC37A2 LRRC37A3
Gene Name:
leucine rich repeat containing 37A
uncharacterized LOC105376839
leucine rich repeat containing 37 member A2
leucine rich repeat containing 37 member A3
Gene Aliases:
LRRC37
LRRC37A
LRRC37
LRRC37, LRRC37A
Chromosome Location:
Chr.17: 46248044 - 46339367 on Build GRCh38
Chromosome Location:
Chr.17: 45588256 - 45598611 on Build GRCh38
Chromosome Location:
Chr.17: 46512657 - 46556747 on Build GRCh38
Chromosome Location:
Chr.17: 64854147 - 64966228 on Build GRCh38
UniGene ID:
Hs.579108
Hs.741051
Hs.646568
Hs.551962
Species:
Human
Species Specific ID (Flybase ID):
-
-
-
-
siRNA ID 241556

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LRRC37A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014834.4 NP_055649.4 1 314
XM_005257879.4 XP_005257936.1 1 572
XM_005257880.4 XP_005257937.1 1 572
XM_005257882.4 XP_005257939.1 2 758
XM_005257883.4 XP_005257940.1 1 572
XM_005257884.4 XP_005257941.1 1 572
XM_006722210.3 XP_006722273.1 1 572
XM_006722211.3 XP_006722274.1 1 572
XM_011525539.2 XP_011523841.1 1 572
XM_017025472.1 XP_016880961.1 3 171
GenBank mRNA BC040501.1 - 1 314 5295808
CR749232.1 - 1 316
Gene Symbol: LOC105376839
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA JQ358763.1 - 6 575
Gene Symbol: LRRC37A2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006607.2 NP_001006608.2 1 314
XM_011524841.2 XP_011523143.1 1 367
XM_011524842.2 XP_011523144.1 1 367
XM_011524843.2 XP_011523145.1 1 367
XM_011524844.2 XP_011523146.1 1 367
XM_011524846.2 XP_011523148.1 1 367
XM_011524848.2 XP_011523150.1 1 367
XM_011524849.1 XP_011523151.1 1 367
XM_011524850.2 XP_011523152.1 1 367
Gene Symbol: LRRC37A3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199340.3 NP_955372.2 4 977
XM_017024593.1 XP_016880082.1 2 806
XM_017024594.1 XP_016880083.1 2 806
XM_017024595.1 XP_016880084.1 2 806
XM_017024596.1 XP_016880085.1 2 806
XM_017024597.1 XP_016880086.1 2 806
XM_017024598.1 XP_016880087.1 2 806
XM_017024599.1 XP_016880088.1 3 178
XM_017024600.1 XP_016880089.1 2 806
GenBank mRNA AB011135.2 - 3 842
BC043145.1 - 1 319 5296147

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-lqf76:80/100.66.77.18:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline