This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other C16ORF52 SIRNA Assays ›
Gene Symbol:
C16orf52
Gene Name:
chromosome 16 open reading frame 52
Gene Aliases:
-
Chromosome Location:
Chr.16: 22008135 - 22084654 on Build GRCh38
UniGene ID:
Hs.498890
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 242020

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C16orf52
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164579.1 NP_001158051.1 2 445
GenBank mRNA BC027604.1 - 3 475
BC053950.1 - 3 462

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006230181.
This siRNA also targets the following transcripts in Mouse: NM_001164580, AK052350, AK162324, BC030336.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-gm6kg:80/100.66.79.106:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0