siRNA Details
| Gene Symbol: TMEM26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178505.7 | NP_848600.2 | 3, 4 | 749 | ||
| NR_134507.1 | 3, 4 | 749 | ||||
| NR_134508.1 | 3, 4 | 749 | ||||
| XM_011539450.2 | XP_011537752.1 | 3, 4 | 537 | |||
| XM_011539451.1 | XP_011537753.1 | 3, 4 | 537 | |||
| XM_017015829.1 | XP_016871318.1 | 3, 4 | 537 | |||
| GenBank mRNA | AK125611.1 | - | 3, 4 | 537 | ||
| BC042872.1 | - | 3, 4 | 466 | 5164050 | ||
| CR749606.1 | - | 3, 4 | 749 | |||

TMEM26
Genomic Map