siRNA Details
| Gene Symbol: SPOUT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016390.3 | NP_057474.2 | 3 | 139 | ||
| XM_017014804.1 | XP_016870293.1 | 4 | 406 | |||
| GenBank mRNA | AF161459.1 | - | 3 | 113 | ||
| AK303563.1 | - | 3 | 129 | |||
| AL110193.1 | - | 3 | 139 | |||
| BC010579.1 | - | 3 | 101 | 4213299 | ||
| BC021273.1 | - | 3 | 112 | 4843384 | ||
| BC033677.1 | - | 3 | 107 | 5176852 | ||
| BC039590.1 | - | 3 | 124 | 5757547 | ||
| BC046133.1 | - | 3 | 104 | 5725479 | ||
| BC063644.1 | - | 3 | 107 | 4539701 | ||

SPOUT1
Genomic Map