siRNA Details
| Gene Symbol: Opa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133585.3 | NP_598269.3 | 19 | 1926 | ||
| XM_006248497.1 | XP_006248559.1 | 21 | 2190 | |||
| XM_006248498.1 | XP_006248560.1 | 20 | 2136 | |||
| XM_006248499.1 | XP_006248561.1 | 20 | 2079 | |||
| GenBank mRNA | AY333988.1 | - | 19 | 1835 | ||
| AY510274.1 | - | 19 | 1981 | |||
| BC111071.1 | - | 19 | 1926 | 7324017 | ||
| U93197.1 | - | 3 | 132 | |||

Opa1
Genomic Map