This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GTF2I SIRNA Assays ›
Gene Symbol:
Gtf2i
Gene Name:
general transcription factor II I
Gene Aliases:
Gtf2ird1
Chromosome Location:
Chr.12: 25410804 - 25487970 on Build Rnor_6.0
UniGene ID:
Rn.27575
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 251568

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gtf2i
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001512.2 NP_001001512.2 31 3063
XM_006249162.2 XP_006249224.1 35 3328
XM_006249163.2 XP_006249225.1 35 3325
XM_006249164.2 XP_006249226.1 34 3268
XM_006249165.2 XP_006249227.1 34 3265
XM_006249166.2 XP_006249228.1 34 3262
XM_006249167.2 XP_006249229.1 33 3211
XM_006249168.2 XP_006249230.1 33 3205
XM_006249169.2 XP_006249231.1 33 3190
XM_006249170.2 XP_006249232.1 35 3523
XM_006249171.2 XP_006249233.1 32 3148
XM_006249172.2 XP_006249234.1 32 3133
XM_006249173.2 XP_006249235.1 32 3127
XM_006249174.2 XP_006249236.1 31 3070
XM_008769141.1 XP_008767363.1 35 3190
GenBank mRNA AF547390.1 - 33 3100
AF547391.1 - 33 3100
BC085815.1 - 31 3201 7123295
DQ294706.1 - 2 159
DQ294707.1 - 2 159

Back To Top

More Information


Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transition between slow and fast fiber
negative regulation of angiogenesis
negative regulation of cytosolic calcium ion concentration
molecular_function
DNA binding
mitogen-activated protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-bcf2z:80/100.66.76.251:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline