This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GTF2I SIRNA Assays ›
Gene Symbol:
Gtf2i
Gene Name:
general transcription factor II I
Gene Aliases:
Gtf2ird1
Chromosome Location:
Chr.12: 25410804 - 25487970 on Build Rnor_6.0
UniGene ID:
Rn.27575
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 251569

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gtf2i
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001512.2 NP_001001512.2 9, 10 954
XM_006249162.2 XP_006249224.1 13, 14 1219
XM_006249163.2 XP_006249225.1 13, 14 1216
XM_006249164.2 XP_006249226.1 12, 13 1159
XM_006249165.2 XP_006249227.1 12, 13 1156
XM_006249166.2 XP_006249228.1 12, 13 1153
XM_006249167.2 XP_006249229.1 11, 12 1102
XM_006249168.2 XP_006249230.1 11, 12 1096
XM_006249169.2 XP_006249231.1 11, 12 1081
XM_006249170.2 XP_006249232.1 13, 14 1414
XM_006249171.2 XP_006249233.1 10, 11 1039
XM_006249172.2 XP_006249234.1 10, 11 1024
XM_006249173.2 XP_006249235.1 10, 11 1018
XM_006249174.2 XP_006249236.1 9, 10 961
XM_008769141.1 XP_008767363.1 13, 14 1081
GenBank mRNA AF547390.1 - 11, 12 991
AF547391.1 - 11, 12 991
BC085815.1 - 9, 10 1092 7123295

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010365, NM_001080746, NM_001080747, NM_001080748, NM_001080749, and others.

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transition between slow and fast fiber
negative regulation of angiogenesis
negative regulation of cytosolic calcium ion concentration
molecular_function
DNA binding
mitogen-activated protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f5rhb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0