siRNA Details
| Gene Symbol: ZNF740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004304.3 | NP_001004304.1 | 7 | 1382 | ||
| XM_017019181.1 | XP_016874670.1 | 7 | 1616 | |||
| GenBank mRNA | AK091946.1 | - | 4 | 765 | ||
| BC053557.1 | - | 7 | 1385 | 6162269 | ||

ZNF740
Genomic Map