This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZYG11A SIRNA Assays ›
Gene Symbol:
ZYG11A
Gene Name:
zyg-11 family member A, cell cycle regulator
Gene Aliases:
ZYG11
Chromosome Location:
Chr.1: 52842511 - 52894575 on Build GRCh38
UniGene ID:
Hs.658458
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 251663

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZYG11A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004339.2 NP_001004339.2 4 1426
NM_001307931.1 NP_001294860.1 3 674
XM_011541480.2 XP_011539782.1 4 1201
XM_011541481.2 XP_011539783.1 4 1201
XM_011541482.2 XP_011539784.1 4 1201
XM_011541483.2 XP_011539785.1 4 1201
XM_011541484.1 XP_011539786.1 4 1201
XM_011541485.1 XP_011539787.1 4 1201
XM_011541486.1 XP_011539788.1 4 1201
XM_011541487.2 XP_011539789.1 4 1201
XM_011541488.1 XP_011539790.1 4 1201
GenBank mRNA AY271826.1 - 3 665

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
regulation of ubiquitin-protein transferase activity
ubiquitin-protein transferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-hssmq:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline