This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LOC100506422 SIRNA Assays ›
Gene Symbol:
LOC100506422
Gene Name:
putative deoxyuridine 5'-triphosphate nucleotidohydrolase-like protein FLJ16323
Gene Aliases:
-
Chromosome Location:
Chr.9: 26066675 - 26126108 on Build GRCh38
UniGene ID:
Hs.533221
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 251673

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC100506422
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004352.2 NP_001004352.1 7 2225
GenBank mRNA AK131322.1 - 7 2226

Back To Top

More Information


Panther Classification:

Molecular Function -

hydrolase phosphatase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

dUTP metabolic process
hydrolase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline