This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ELP6 SIRNA Assays ›
Gene Symbol:
ELP6
Gene Name:
elongator acetyltransferase complex subunit 6
Gene Aliases:
C3orf75, TMEM103
Chromosome Location:
Chr.3: 47495640 - 47513711 on Build GRCh38
UniGene ID:
Hs.311100
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 25206

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ELP6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031703.2 NP_001026873.2 5 454
XM_005265241.3 XP_005265298.1 4 380
XM_011533851.2 XP_011532153.1 5 456
XM_011533852.2 XP_011532154.1 5 792
XM_011533853.2 XP_011532155.1 5 433
XM_011533854.2 XP_011532156.1 4 357
XM_011533856.2 XP_011532158.1 4 412
XM_011533857.2 XP_011532159.1 4 582
XM_017006669.1 XP_016862158.1 5 456
XM_017006670.1 XP_016862159.1 5 725
GenBank mRNA AK000218.1 - 5 439
AK225721.1 - 5 436
AK298202.1 - 4 389
BC000623.2 - 5 358 3346325
BC014204.2 - 5 358 4620013
DQ787088.1 - 5 335

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
biological_process
positive regulation of cell migration
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-s92gb:80/100.66.74.196:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0