This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other BAG3 SIRNA Assays ›
Gene Symbol:
Bag3
Gene Name:
Bcl2-associated athanogene 3
Gene Aliases:
-
Chromosome Location:
Chr.1: 199941258 - 199965191 on Build Rnor_6.0
UniGene ID:
Rn.46304
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 252092

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Bag3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011936.1 NP_001011936.1 4 2226
GenBank mRNA BC085857.1 - 4 2226 7190249

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

brain development
extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of striated muscle cell apoptotic process
spinal cord development
protein stabilization
cellular response to mechanical stimulus
extrinsic apoptotic signaling pathway in absence of ligand
protein complex binding
chaperone binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-fbxsq:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline