siRNA Details
| Gene Symbol: Rgs7bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012347.1 | NP_001012347.1 | 1176 | |||
| GenBank mRNA | BC089977.1 | - | 1176 | 7317427 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rgs7bp| Gene Symbol: Rgs7bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012347.1 | NP_001012347.1 | 1176 | |||
| GenBank mRNA | BC089977.1 | - | 1176 | 7317427 | ||