This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RGS7BP SIRNA Assays ›
Gene Symbol:
Rgs7bp
Gene Name:
regulator of G-protein signaling 7 binding protein
Gene Aliases:
R7bp, RGD1309360
Chromosome Location:
Chr.2: 35077300 - 35104743 on Build Rnor_6.0
UniGene ID:
Rn.198108
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 252286

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rgs7bp
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012347.1 NP_001012347.1 1111
GenBank mRNA BC089977.1 - 1111 7317427

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029879, BC118957, AK028323, AK137417, AK043529, and others.

Gene Ontology Categories:

Function(s)

G-protein coupled receptor signaling pathway
negative regulation of signal transduction

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-n4nrd:80/100.66.76.224:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline